Gene Transcripts

Gene Symbol
Lama3
Entrez Gene ID
16774
Gene Name
laminin, alpha 3
Gene Aliases
Lama3B, [a]3B
Chromosome Location
Chr. 18: 12333723 - 12583013 on Build GRCm38
UniGene ID
Mm.42012
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010680.1 NP_034810.1 62 8064
GenBank mRNA AK141255.1 - 7 777
X84013.1 - 25 3270
X84014.1 - 43 5782

View Full Product Details