Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001561.5 | NP_001552.2 | 4, 5 | 477 | |
XM_006710618.3 | XP_006710681.1 | 3, 4 | 1124 | ||
XM_011541386.2 | XP_011539688.1 | 3, 4 | 492 | ||
GenBank mRNA | BC006196.2 | - | 3, 4 | 337 | |
BP381234.1 | - | 4, 5 | 477 | ||
L12964.2 | - | 3, 4 | 355 | ||
U03397.1 | - | 3, 4 | 335 |