Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023178.2 | NP_075667.1 | 3 | 359 | |
GenBank mRNA | AB221661.1 | - | 3 | 302 | |
AF265229.1 | - | 3 | 302 | ||
AF438610.1 | - | 3 | 359 | ||
AK005270.1 | - | 4 | 399 | ||
AK012055.1 | - | 4 | 380 | ||
AK136976.1 | - | 2 | 592 | ||
BC002321.1 | - | 3 | 252 | ||
BC045160.1 | - | 3 | 339 | 6309705 |