Gene Transcripts

Gene Symbol
Ccdc105
Entrez Gene ID
70976
Gene Name
coiled-coil domain containing 105
Gene Aliases
4931413A09Rik
Chromosome Location
Chr. 10: 78746924 - 78753067 on Build GRCm38
UniGene ID
Mm.188505
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027630.1 NP_081906.1 5 1203
GenBank mRNA AK016452.1 - 5 1204

View Full Product Details