Gene Transcripts

Gene Symbol
1700074P13Rik
Entrez Gene ID
73481
Gene Name
RIKEN cDNA 1700074P13 gene
Gene Aliases
AV047902, Tryx5
Chromosome Location
Chr. 6: 40920437 - 40940973 on Build GRCm38
UniGene ID
Mm.333010
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028550.3 NP_082826.1 6 941
GenBank mRNA AK006951.1 - 5 815
AK030264.1 - 6 941
BC061035.1 - 5 852 6774253

View Full Product Details