gRNA Design Details
- Target DNA Sequence
- GCAGGCCCTACTATTCCTTA
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 56062250 - 56062272 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.93/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mcpt4
- Entrez Gene ID
-
17227
- Gene Name
- mast cell protease 4
- Gene Aliases
-
MMCP-4, MMCP-4A, MMCP-4B, Mcp-4, Mcp4
- Chromosome Location
-
Chr.14: 56059744 - 56062310 on GRCm38
- UniGene ID
- Mm.439684
- Species
-
Mouse