gRNA Design Details
- Target DNA Sequence
- CACCACTAAGAACGGTTCGA
- PAM Sequence
- AGG
- Target locus
-
Chr.14: 56042765 - 56042787 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mcpt2
- Entrez Gene ID
-
17225
- Gene Name
- mast cell protease 2
- Gene Aliases
-
MMCP-2, Mcp-2
- Chromosome Location
-
Chr.14: 56021584 - 56044635 on GRCm38
- UniGene ID
- Mm.4409
- Species
-
Mouse