Gene Transcripts

Gene Symbol
Vmn1r21
Entrez Gene ID
171201
Gene Name
vomeronasal 1 receptor 21
Gene Aliases
V1rc28
Chromosome Location
Chr. 6: 57843564 - 57844457 on Build GRCm38
UniGene ID
Mm.377179
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134183.1 NP_598944.1 1 325
GenBank mRNA BC120799.1 - 1 366
BC120801.1 - 1 366 40129724

View Full Product Details