Gene Transcripts

Gene Symbol
Olfr541
Entrez Gene ID
258964
Gene Name
olfactory receptor 541
Gene Aliases
MOR253-3
Chromosome Location
Chr. 7: 140704253 - 140705191 on Build GRCm38
UniGene ID
Mm.389986
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146962.1 NP_667173.1 1 922
GenBank mRNA BC119150.1 - 1 1010
BC120836.1 - 1 1010 40129759

View Full Product Details