Gene Transcripts

Gene Symbol
USP19
Entrez Gene ID
10869
Gene Name
ubiquitin specific peptidase 19
Gene Aliases
ZMYND9
Chromosome Location
Chr. 3: 49108046 - 49120938 on Build GRCh38
UniGene ID
Hs.255596
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199160.1 NP_001186089.1 2 408
NM_001199161.1 NP_001186090.1 2 408
NM_001199162.1 NP_001186091.1 2 408
NM_006677.2 NP_006668.1 2 408
XM_005264823.2 XP_005264880.1 2 403
XM_005264824.2 XP_005264881.1 2 422
XM_005264825.2 XP_005264882.1 2 403
XM_005264826.2 XP_005264883.1 2 403
XM_005264827.2 XP_005264884.1 2 403
XM_005264829.2 XP_005264886.1 2 403
XM_005264830.2 XP_005264887.1 2 403
XM_005264831.2 XP_005264888.1 2 403
XM_006712946.2 XP_006713009.1 2 403
XM_006712947.2 XP_006713010.1 2 403
XM_006712948.2 XP_006713011.1 2 403
XM_006712949.2 XP_006713012.1 2 403
XM_006712950.2 XP_006713013.1 2 403
XM_006712951.2 XP_006713014.1 2 403
XM_006712952.2 XP_006713015.1 2 403
XM_006712953.1 XP_006713016.1 2 374
XM_017005615.1 XP_016861104.1 2 403
XM_017005616.1 XP_016861105.1 2 403
XM_017005617.1 XP_016861106.1 2 403
XM_017005618.1 XP_016861107.1 2 403
XM_017005619.1 XP_016861108.1 2 403
XM_017005620.1 XP_016861109.1 2 403
XM_017005621.1 XP_016861110.1 2 403
XM_017005622.1 XP_016861111.1 2 403
XM_017005623.1 XP_016861112.1 2 403
XM_017005624.1 XP_016861113.1 2 403
XM_017005625.1 XP_016861114.1 2 403
XM_017005626.1 XP_016861115.1 2 422
XM_017005627.1 XP_016861116.1 2 403
XM_017005628.1 XP_016861117.1 2 403
XM_017005629.1 XP_016861118.1 2 524
XM_017005630.1 XP_016861119.1 2 403
XM_017005631.1 XP_016861120.1 2 403
XM_017005632.1 XP_016861121.1 2 403
XM_017005633.1 XP_016861122.1 2 403
XM_017005634.1 XP_016861123.1 2 403
GenBank mRNA AB020698.1 - 2 250
AK294756.1 - 2 301
AK296447.1 - 2 288
AK300425.1 - 2 262
BC106029.1 - 2 271
BC142660.1 - 2 278
BC142727.1 - 2 284
BC146752.1 - 2 250 9007186
DC323791.1 - 2 408
DC332671.1 - 2 373

View Full Product Details