Gene Transcripts

Gene Symbol
SPOCD1
Entrez Gene ID
90853
Gene Name
SPOC domain containing 1
Gene Aliases
PPP1R146
Chromosome Location
Chr. 1: 31790422 - 31816051 on Build GRCh38
UniGene ID
Hs.62604
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281987.1 NP_001268916.1 12 2603
NM_001281988.1 NP_001268917.1 11 1181
NM_144569.5 NP_653170.3 12 2603
XM_011542413.2 XP_011540715.1 12 1172
XM_011542414.2 XP_011540716.1 11 1367
XM_017002779.1 XP_016858268.1 11 2509
XM_017002780.1 XP_016858269.1 10 2470
XM_017002781.1 XP_016858270.1 10 2386
XM_017002782.1 XP_016858271.1 11 1741
GenBank mRNA AK058077.1 - 2 183
AK091016.1 - 11 1152
AK131447.1 - 12 2532
BC029069.1 - 4 493
BC114430.1 - 12 2529 40038407

View Full Product Details