Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020165.3 | NP_064550.3 | 13 | 1560 | |
XM_017006873.1 | XP_016862362.1 | 14 | 1718 | ||
GenBank mRNA | AB035274.2 | - | 13 | 1595 | |
AF169796.1 | - | 13 | 1541 | ||
AK023075.1 | - | 13 | 1547 | ||
AK222771.1 | - | 13 | 1538 | ||
AY004333.1 | - | 13 | 1506 | ||
BC001302.1 | - | 13 | 1527 | 3451960 |