Gene Transcripts
- Gene Symbol
- PDPK1
- Entrez Gene ID
- 5170
- Gene Name
- 3-phosphoinositide dependent protein kinase 1
- Gene Aliases
- PDK1, PDPK2, PDPK2P, PRO0461
- Chromosome Location
- Chr. 16: 2537964 - 2603190 on Build GRCh38
- UniGene ID
- Hs.459691
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001261816.1 |
NP_001248745.1
|
4, 5 |
619 |
|
NM_002613.4 |
NP_002604.1
|
4, 5 |
619 |
|
XM_011522521.2 |
XP_011520823.1
|
4, 5 |
4307 |
|
XM_017023271.1 |
XP_016878760.1
|
4, 5 |
4312 |
|
XM_017023272.1 |
XP_016878761.1
|
4, 5 |
868 |
|
XM_017023273.1 |
XP_016878762.1
|
4, 5 |
577 |
|
XM_017023274.1 |
XP_016878763.1
|
5, 6 |
715 |
|
GenBank mRNA |
AB209835.1 |
- |
4, 5 |
586 |
|
AF017995.1 |
- |
4, 5 |
550 |
|
AJ785968.1 |
- |
3, 4 |
1253 |
|
AK131060.1 |
- |
4, 5 |
553 |
|
AK222581.1 |
- |
4, 5 |
562 |
|
BC012103.1 |
- |
4, 5 |
579 |
4561371 |
CR536517.1 |
- |
4, 5 |
470 |
|
View Full Product Details