Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153563.2 | NP_705791.2 | 7 | 766 | |
GenBank mRNA | AK032065.1 | - | 7 | 748 | |
AK035833.1 | - | 7 | 751 | ||
AK122553.1 | - | 7 | 730 | ||
AK148011.1 | - | 7 | 751 | ||
AK169644.1 | - | 7 | 698 | ||
AK178114.1 | - | 2 | 146 | ||
AK187262.1 | - | 2 | 146 | ||
BC023952.1 | - | 7 | 742 | 5320305 |