Gene Transcripts

Gene Symbol
Plekha3
Entrez Gene ID
83435
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 3
Gene Aliases
FAPP1
Chromosome Location
Chr. 2: 76675310 - 76697335 on Build GRCm38
UniGene ID
Mm.41636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031256.3 NP_112546.1 3 565
GenBank mRNA AF286163.1 - 3 354
BC012283.1 - 3 305
BC031110.1 - 4 373 4981486
BY730332.1 - 3 565

View Full Product Details