Gene Transcripts

Gene Symbol
Mesdc2
Entrez Gene ID
67943
Gene Name
mesoderm development candidate 2
Gene Aliases
2210015O11Rik, AW537813, mKIAA0081, mesd, msd
Chromosome Location
Chr. 7: 83892000 - 83901532 on Build GRCm38
UniGene ID
Mm.117365
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023403.3 NP_075892.3 3 3066
GenBank mRNA AK031949.1 - 3 2809
BC058612.1 - 5720

View Full Product Details