Gene Transcripts

Gene Symbol
Kctd2
Entrez Gene ID
70382
Gene Name
potassium channel tetramerisation domain containing 2
Gene Aliases
2310012I15Rik
Chromosome Location
Chr. 11: 115420126 - 115431274 on Build GRCm38
UniGene ID
Mm.276299
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183285.3 NP_899108.3 3 554
GenBank mRNA AA537633.1 - 3 208
AK009318.1 - 3 551
BC095988.1 - 3 513 5365458

View Full Product Details