Gene Transcripts

Gene Symbol
Cxcl11
Entrez Gene ID
305236
Gene Name
chemokine (C-X-C motif) ligand 11
Gene Aliases
SCYB11
Chromosome Location
Chr. 14: 17195395 - 17198170 on Build Rnor_6.0
UniGene ID
Rn.13664
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182952.2 NP_891997.1 2 199
GenBank mRNA AF537209.1 - 2 199
AY340181.1 - 2 178
BC168725.1 - 2 207 6919932

View Full Product Details