Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053541.1 | NP_445993.1 | 7 | 3288 | |
XM_008759197.1 | XP_008757419.1 | 6 | 4568 | ||
XM_008759200.1 | XP_008757422.1 | 7 | 3487 | ||
XM_008759201.1 | XP_008757423.1 | 6 | 4447 | ||
XM_008759202.1 | XP_008757424.1 | 7 | 3367 | ||
GenBank mRNA | AB009463.1 | - | 7 | 3288 |