Gene Transcripts

Gene Symbol
OIP5
Entrez Gene ID
11339
Gene Name
Opa interacting protein 5
Gene Aliases
5730547N13Rik, CT86, LINT-25, MIS18B, MIS18beta, hMIS18beta
Chromosome Location
Chr. 15: 41309268 - 41332621 on Build GRCh38
UniGene ID
Hs.661645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007280.1 NP_009211.1 2, 3 441
GenBank mRNA AF025441.1 - 2, 3 387
BC015050.1 - 2, 3 441 3908972

View Full Product Details