Gene Transcripts

Gene Symbol
Nefm
Entrez Gene ID
24588
Gene Name
neurofilament, medium polypeptide
Gene Aliases
Nef3, Nfm
Chromosome Location
Chr. 15: 44855307 - 44860647 on Build Rnor_6.0
UniGene ID
Rn.10971
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017029.2 NP_058725.1 3 1255
XM_008770736.1 XP_008768958.1 4 1592
GenBank mRNA M18628.1 - 3 1233
Z12152.1 - 3 1258

View Full Product Details