Gene Transcripts

Gene Symbol
Pgr
Entrez Gene ID
25154
Gene Name
progesterone receptor
Gene Aliases
Chromosome Location
Chr. 8: 7128656 - 7187796 on Build Rnor_6.0
UniGene ID
Rn.10303
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022847.1 NP_074038.1 1 215
GenBank mRNA HM037361.1 - 758
HM037362.1 - 758
L16922.1 - 1 215

View Full Product Details