Gene Transcripts
- Gene Symbol
- Dbi
- Entrez Gene ID
- 25045
- Gene Name
- diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein)
- Gene Aliases
- Acbp, Acoabp3, Ep, Odn, RNACOABP3, Ttn
- Chromosome Location
- Chr. 13: 36147667 - 36156076 on Build Rnor_6.0
- UniGene ID
- Rn.3285
- Species
- Rattus norvegicus
- Gene Symbol
- LOC301444
- Entrez Gene ID
- 301444
- Gene Name
- pseudogene for diazepam binding inhibitor 1
- Gene Aliases
- Ac1-130
- Chromosome Location
- Chr. 9: 66712547 - 66712874 on Build Rnor_6.0
- UniGene ID
- Rn.217593
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY321319.1 |
- |
1 |
137 |
|
View Full Product Details