Gene Transcripts

Gene Symbol
Gfpt2
Entrez Gene ID
360518
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Gene Aliases
Chromosome Location
Chr. 10: 35279237 - 35325506 on Build Rnor_6.0
UniGene ID
Rn.16089
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002819.2 NP_001002819.2 5 489
GenBank mRNA BC098630.1 - 5 489

View Full Product Details