Gene Transcripts
- Gene Symbol
- Ryk
- Entrez Gene ID
- 20187
- Gene Name
- receptor-like tyrosine kinase
- Gene Aliases
- AW536699, ERK-3, Vik
- Chromosome Location
- Chr. 9: 102834920 - 102908307 on Build GRCm38
- UniGene ID
- Mm.335391
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042607.1 |
NP_001036072.1
|
11, 12 |
1387 |
|
NM_001284258.1 |
NP_001271187.1
|
10, 11 |
1265 |
|
NM_013649.3 |
NP_038677.3
|
11, 12 |
1396 |
|
GenBank mRNA |
AK076449.1 |
- |
5, 6 |
489 |
|
AK144355.1 |
- |
10, 11 |
1205 |
|
BC006963.1 |
- |
7, 8 |
695 |
|
BC032275.1 |
- |
11, 12 |
1121 |
5370100 |
L02210.1 |
- |
11, 12 |
1371 |
|
L21707.1 |
- |
11, 12 |
1405 |
|
L38394.1 |
- |
11, 12 |
1319 |
|
M98547.1 |
- |
11, 12 |
1360 |
|
View Full Product Details