Gene Transcripts
- Gene Symbol
- Usp19
- Entrez Gene ID
- 71472
- Gene Name
- ubiquitin specific peptidase 19
- Gene Aliases
- 8430421I07Rik, AI047774, Zmynd9
- Chromosome Location
- Chr. 9: 108490607 - 108502337 on Build GRCm38
- UniGene ID
- Mm.289706
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001168371.2 |
NP_001161843.1
|
3, 4 |
509 |
|
NM_001168372.2 |
NP_001161844.1
|
3, 4 |
509 |
|
NM_001168373.2 |
NP_001161845.1
|
3, 4 |
509 |
|
NM_027804.4 |
NP_082080.3
|
3, 4 |
509 |
|
NM_145407.3 |
NP_663382.2
|
3, 4 |
509 |
|
GenBank mRNA |
AK043088.1 |
- |
3, 4 |
345 |
|
AK122396.2 |
- |
2, 3 |
295 |
|
AK146504.1 |
- |
3, 4 |
335 |
|
AK159473.1 |
- |
3, 4 |
330 |
|
AK160807.1 |
- |
3, 4 |
335 |
|
AK171942.1 |
- |
3, 4 |
341 |
|
BC060613.1 |
- |
3, 4 |
437 |
6400986 |
BQ442290.1 |
- |
2, 3 |
297 |
|
CN665283.1 |
- |
3, 4 |
496 |
|
View Full Product Details