Gene Transcripts
- Gene Symbol
- CRISP1
- Entrez Gene ID
- 167
- Gene Name
- cysteine rich secretory protein 1
- Gene Aliases
- AEGL1, ARP, CRISP-1, HEL-S-57, HSCRISP1D, HSCRISP1G, HUMARP
- Chromosome Location
- Chr. 6: 49834257 - 49877096 on Build GRCh38
- UniGene ID
- Hs.109620
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001131.2 |
NP_001122.2
|
2, 3 |
156 |
|
NM_001205220.1 |
NP_001192149.1
|
2, 3 |
167 |
|
NM_170609.1 |
NP_733758.1
|
2, 3 |
156 |
|
GenBank mRNA |
AB451334.1 |
- |
1, 2 |
77 |
|
AB451479.1 |
- |
1, 2 |
77 |
|
AK292497.1 |
- |
2, 3 |
137 |
|
BC131707.1 |
- |
2, 3 |
127 |
|
D38451.1 |
- |
2, 4 |
156 |
|
GQ472203.1 |
- |
2, 3 |
150 |
|
S80310.1 |
- |
2, 4 |
156 |
|
X95237.1 |
- |
2, 3 |
150 |
|
X95238.1 |
- |
2, 3 |
150 |
|
View Full Product Details