Gene Transcripts

Gene Symbol
ZNF160
Entrez Gene ID
90338
Gene Name
zinc finger protein 160
Gene Aliases
F11, HKr18, HZF5, KR18
Chromosome Location
Chr. 19: 53064882 - 53103434 on Build GRCh38
UniGene ID
Hs.655967
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102603.1 NP_001096073.1 1 113
NM_001322125.1 NP_001309054.1 1 113
NM_001322126.1 NP_001309055.1 1 113
NM_001322128.1 NP_001309057.1 1 113
NM_001322129.1 NP_001309058.1 1 113
NM_001322130.1 NP_001309059.1 1 113
NM_001322131.1 NP_001309060.1 1 113
NM_001322132.1 NP_001309061.1 1 113
NM_001322133.1 NP_001309062.1 1 113
NM_001322134.1 NP_001309063.1 1 113
NM_001322135.1 NP_001309064.1 1 113
NM_001322137.1 NP_001309066.1 1 113
NM_001322138.1 NP_001309067.1 1 113
NM_001322139.1 NP_001309068.1 1 113
NM_033288.3 NP_150630.2 1 113
NM_198893.2 NP_942596.1 1 113
GenBank mRNA AF277623.1 - 1 121
BC000807.1 - 1 82
BC094880.1 - 1 30 3066532
BU430880.1 - 1 37
DA230397.1 - 1 175
DB078640.1 - 1 113

View Full Product Details