Gene Transcripts

Gene Symbol
Olr1171
Entrez Gene ID
405373
Gene Name
olfactory receptor 1171
Gene Aliases
Chromosome Location
Chr. 8: 20397606 - 20398544 on Build Rnor_6.0
UniGene ID
Rn.143581
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001005.1 NP_001001005.1 1 13

View Full Product Details