Gene Transcripts

Gene Symbol
ZNF275
Entrez Gene ID
10838
Gene Name
zinc finger protein 275
Gene Aliases
Chromosome Location
Chr. X: 153334155 - 153352926 on Build GRCh38
UniGene ID
Hs.348963
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080485.3 NP_001073954.3 5 2118
GenBank mRNA AK122976.1 - 5 2118
AK127133.1 - 1 1914
BC136634.1 - 2 3242 9020624

View Full Product Details