Gene Transcripts

Gene Symbol
LOC439933
Entrez Gene ID
439933
Gene Name
uncharacterized LOC439933
Gene Aliases
Chromosome Location
Chr. 4: 36244116 - 36274215 on Build GRCh38
UniGene ID
Hs.591071
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_122079.1 5 693
NR_122080.1 3 568
GenBank mRNA BC030111.1 - 3 574
BC037904.2 - 5 693

View Full Product Details