Gene Transcripts
- Gene Symbol
- ZNF317
- Entrez Gene ID
- 57693
- Gene Name
- zinc finger protein 317
- Gene Aliases
- Chromosome Location
- Chr. 19: 9140380 - 9163419 on Build GRCh38
- UniGene ID
- Hs.465829
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190791.1 |
NP_001177720.1
|
3, 4 |
475 |
|
NM_020933.4 |
NP_065984.3
|
3, 4 |
475 |
|
NR_102435.1 |
|
3, 4 |
475 |
|
GenBank mRNA |
AB046808.1 |
- |
2, 3 |
252 |
|
AF148135.1 |
- |
3, 4 |
458 |
|
AF275255.1 |
- |
3, 4 |
458 |
|
AF307096.1 |
- |
2 |
692 |
|
AF307097.1 |
- |
2 |
694 |
|
AK056570.1 |
- |
3, 4 |
458 |
|
AK304151.1 |
- |
2, 3 |
309 |
|
AK304871.1 |
- |
3, 4 |
427 |
|
AK314560.1 |
- |
3, 4 |
458 |
|
AL834126.2 |
- |
3, 4 |
436 |
|
BC078154.1 |
- |
3, 4 |
445 |
6060418 |
DA757943.1 |
- |
3, 4 |
475 |
|
View Full Product Details