Gene Transcripts

Gene Symbol
Olfr1383
Entrez Gene ID
404337
Gene Name
olfactory receptor 1383
Gene Aliases
MOR256-56
Chromosome Location
Chr. 11: 49523725 - 49524660 on Build GRCm38
UniGene ID
Mm.278373
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207574.1 NP_997457.1 1 148
GenBank mRNA BC127965.1 - 1 152

View Full Product Details