Gene Transcripts

Gene Symbol
C15orf32
Entrez Gene ID
145858
Gene Name
chromosome 15 open reading frame 32
Gene Aliases
Chromosome Location
Chr. 15: 92465762 - 92501117 on Build GRCh38
UniGene ID
Hs.367879
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153040.2 NP_694585.1 2, 3 969
GenBank mRNA AK057393.1 - 2, 3 970
BC109253.1 - 2, 3 723 40023844

View Full Product Details