Gene Transcripts

Gene Symbol
Klk13
Entrez Gene ID
626834
Gene Name
kallikrein related-peptidase 13
Gene Aliases
Egfbp-2, mGk-13
Chromosome Location
Chr. 7: 43712567 - 43727864 on Build GRCm38
UniGene ID
Mm.389891
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039042.2 NP_001034131.2 6 931
GenBank mRNA BC063763.1 - 6 932

View Full Product Details