Gene Transcripts

Gene Symbol
Smcp
Entrez Gene ID
24899
Gene Name
sperm mitochondria-associated cysteine-rich protein
Gene Aliases
Mcs, Mcsp
Chromosome Location
Chr. 2: 192775425 - 192780631 on Build Rnor_6.0
UniGene ID
Rn.10492
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031536.2 NP_113724.1 2 591
GenBank mRNA BC078795.1 - 2 502
U48702.1 - 2 597
X87883.1 - 2 583

View Full Product Details