Gene Transcripts

Gene Symbol
Rab38
Entrez Gene ID
252916
Gene Name
RAB38, member RAS oncogene family
Gene Aliases
R, Ruby
Chromosome Location
Chr. 1: 152072716 - 152153449 on Build Rnor_6.0
UniGene ID
Rn.9824
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145774.2 NP_665717.1 2 448
GenBank mRNA BC070513.1 - 2 458
M94043.1 - 2 451

View Full Product Details