Gene Transcripts
- Gene Symbol
- Tnnt2
- Entrez Gene ID
- 21956
- Gene Name
- troponin T2, cardiac
- Gene Aliases
- Tnt, cTnT
- Chromosome Location
- Chr. 1: 135836316 - 135852268 on Build GRCm38
- UniGene ID
- Mm.247470
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130174.2 |
NP_001123646.1
|
6, 7 |
303 |
|
NM_001130175.2 |
NP_001123647.4
|
6, 7 |
299 |
|
NM_001130176.1 |
NP_001123648.1
|
4, 5 |
190 |
|
NM_001130177.2 |
NP_001123649.1
|
5, 6 |
291 |
|
NM_001130178.2 |
NP_001123650.1
|
5, 6 |
273 |
|
NM_001130179.2 |
NP_001123651.4
|
5, 6 |
269 |
|
NM_001130180.2 |
NP_001123652.1
|
4, 5 |
261 |
|
NM_001130181.2 |
NP_001123653.4
|
4, 5 |
257 |
|
NM_011619.3 |
NP_035749.1
|
4, 5 |
261 |
|
GenBank mRNA |
AK146945.1 |
- |
6, 7 |
248 |
|
AK163786.1 |
- |
5, 6 |
236 |
|
AK168393.1 |
- |
5, 6 |
236 |
|
AK169061.1 |
- |
5, 6 |
218 |
|
BC063753.1 |
- |
6, 7 |
279 |
30293959 |
BQ554639.1 |
- |
5, 6 |
209 |
|
L47549.1 |
- |
5, 6 |
187 |
|
L47550.1 |
- |
5, 6 |
172 |
|
L47551.1 |
- |
4, 5 |
173 |
|
L47552.1 |
- |
4, 5 |
182 |
|
L47553.1 |
- |
4, 5 |
189 |
|
L47599.1 |
- |
5, 6 |
193 |
|
L47600.1 |
- |
4, 5 |
190 |
|
View Full Product Details