Gene Transcripts

Gene Symbol
Mfsd3
Entrez Gene ID
69572
Gene Name
major facilitator superfamily domain containing 3
Gene Aliases
2310010G13Rik
Chromosome Location
Chr. 15: 76701542 - 76704239 on Build GRCm38
UniGene ID
Mm.34920
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027122.3 NP_081398.2 5 1469
GenBank mRNA AK009269.1 - 5 1548
AK089539.1 - 5 1470
BC019171.1 - 5 1496 5003354

View Full Product Details