Gene Transcripts

Gene Symbol
Anln
Entrez Gene ID
68743
Gene Name
anillin, actin binding protein
Gene Aliases
1110037A17Rik, 2900037I21Rik, C78101, Scraps
Chromosome Location
Chr. 9: 22331214 - 22389696 on Build GRCm38
UniGene ID
Mm.282751
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028390.3 NP_082666.2 4 758
GenBank mRNA BC027141.1 - 4 722
BC032164.1 - 4 776 5346703

View Full Product Details