Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000364.3 | NP_000355.2 | 10 | 451 | |
NM_001001430.2 | NP_001001430.1 | 9 | 421 | ||
NM_001001431.2 | NP_001001431.1 | 9 | 421 | ||
NM_001001432.2 | NP_001001432.1 | 8 | 406 | ||
NM_001276345.1 | NP_001263274.1 | 10 | 451 | ||
NM_001276347.1 | NP_001263276.1 | 9 | 554 | ||
XM_006711508.3 | XP_006711571.1 | 9 | 451 | ||
XM_006711509.3 | XP_006711572.1 | 9 | 546 | ||
XM_011509938.2 | XP_011508240.1 | 10 | 483 | ||
XM_011509939.1 | XP_011508241.1 | 10 | 495 | ||
XM_011509940.2 | XP_011508242.1 | 10 | 499 | ||
XM_011509941.2 | XP_011508243.1 | 10 | 496 | ||
XM_011509942.2 | XP_011508244.1 | 8 | 453 | ||
XM_011509943.2 | XP_011508245.1 | 8 | 437 | ||
XM_011509944.2 | XP_011508246.1 | 8 | 450 | ||
XM_011509946.1 | XP_011508248.1 | 9 | 524 | ||
XM_017002216.1 | XP_016857705.1 | 9 | 429 | ||
XM_017002217.1 | XP_016857706.1 | 9 | 614 | ||
GenBank mRNA | AJ709280.1 | - | 9 | 441 | |
AK125236.1 | - | 8 | 1568 | ||
AK290621.1 | - | 9 | 616 | ||
AK309493.1 | - | 3 | 255 | ||
AY277394.1 | - | 7 | 364 | ||
BC002653.2 | - | 9 | 387 | 3612968 | |
L40162.1 | - | 9 | 382 | ||
S64668.1 | - | 8 | 341 | ||
X74819.1 | - | 8 | 379 | ||
X79856.1 | - | 9 | 359 | ||
X79859.1 | - | 7 | 583 | ||
X79861.1 | - | 8 | 371 |