Gene Transcripts

Gene Symbol
B4GALNT1
Entrez Gene ID
2583
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 1
Gene Aliases
GALGT, GALNACT, GalNAc-T, SPG26
Chromosome Location
Chr. 12: 57623409 - 57633239 on Build GRCh38
UniGene ID
Hs.159481
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276468.1 NP_001263397.1 2 347
NM_001276469.1 NP_001263398.1 2 674
NM_001478.4 NP_001469.1 2 674
XM_005268773.4 XP_005268830.1 2 634
XM_011538147.2 XP_011536449.1 2 634
XM_011538148.2 XP_011536450.1 2 634
XM_017019140.1 XP_016874629.1 2 634
XM_017019141.1 XP_016874630.1 2 346
XM_017019142.1 XP_016874631.1 2 346
GenBank mRNA AB209460.1 - 2 586
AK289690.1 - 2 627
AK293432.1 - 2 313
AK299845.1 - 2 674
AK302503.1 - 1 753
BC029828.1 - 2 637 5176204
DA483728.1 - 2 348
M83651.1 - 2 265

View Full Product Details