Gene Transcripts

Gene Symbol
RPS4Y2
Entrez Gene ID
140032
Gene Name
ribosomal protein S4, Y-linked 2
Gene Aliases
RPS4Y2P
Chromosome Location
Chr. Y: 20756068 - 20781032 on Build GRCh38
UniGene ID
Hs.367761
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039567.2 NP_001034656.1 7 832
GenBank mRNA AF497481.2 - 7 832

View Full Product Details