Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002986.2 | NP_002977.1 | 3 | 390 | |
GenBank mRNA | BC017850.1 | - | 3 | 316 | |
BF197516.1 | - | 1 | 63 | ||
CR457421.1 | - | 3 | 249 | ||
U46573.1 | - | 3 | 302 | ||
Z75668.1 | - | 4 | 304 |