Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_148178.2 | NP_680544.1 | 2 | 177 | |
NM_148179.2 | NP_680545.1 | 2 | 381 | ||
GenBank mRNA | AL528391.3 | - | 2 | 177 | |
BC032136.2 | - | 2 | 143 | 4869926 | |
BQ224958.1 | - | 2 | 345 | ||
DT218980.1 | - | 2 | 349 |