Gene Transcripts

Gene Symbol
MAGI2-AS3
Entrez Gene ID
100505881
Gene Name
MAGI2 antisense RNA 3
Gene Aliases
Chromosome Location
Chr. 7: 79452957 - 79471208 on Build GRCh38
UniGene ID
Hs.31474
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_038343.2 1, 2 283
NR_038344.1 1, 2 283
GenBank mRNA AK311103.1 - 1, 2 251
BI521274.1 - 1, 2 283
CN311982.1 - 1, 2 250

View Full Product Details