Gene Transcripts
- Gene Symbol
- LOC100130920
- Entrez Gene ID
- 100130920
- Gene Name
- uncharacterized LOC100130920
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.683904
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093639.1 |
- |
1 |
692 |
|
- Gene Symbol
- PRKCQ
- Entrez Gene ID
- 5588
- Gene Name
- protein kinase C theta
- Gene Aliases
- PRKCT, nPKC-theta
- Chromosome Location
- Chr. 10: 6393038 - 6580646 on Build GRCh38
- UniGene ID
- Hs.498570
- Species
- Homo sapiens
View Full Product Details