Gene Transcripts

Gene Symbol
CNEP1R1
Entrez Gene ID
255919
Gene Name
CTD nuclear envelope phosphatase 1 regulatory subunit 1
Gene Aliases
C16orf69, NEP1-R1, NEP1R1, TMEM188, TMP125
Chromosome Location
Chr. 16: 50025206 - 50037088 on Build GRCh38
UniGene ID
Hs.59134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281789.1 NP_001268718.1 2, 3 206
NM_153261.5 NP_694993.2 3, 4 257
GenBank mRNA AK095420.1 - 2, 3 179
AK301610.1 - 3, 4 222
AK307132.1 - 2, 3 253
BC036683.2 - 3, 4 186 5266084
CR933600.1 - 3, 4 202
FJ516381.1 - 2, 3 134
HY011229.1 - 2, 3 207

View Full Product Details