Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72110218-72110719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC15
Entrez Gene ID
25906
Gene Name
anaphase promoting complex subunit 15
Gene Aliases
APC15, C11orf51, HSPC020
UniGene
Hs.38044
Gene Chromosome Location
Chr.11: 72106254-72112793 on Build GRCh38
Transcripts
NM_001278485.1 NM_001278486.1 NM_001278487.1 NM_001278488.1 NM_001278489.1 NM_001278490.1 NM_001278491.1 NM_001278492.1 NM_001278493.1 NM_001278494.1 NM_014042.2 XM_017017488.1 XM_017017489.1 XM_017017490.1 XM_017017491.1 XM_017017492.1 XM_017017493.1 XM_017017494.1 XM_017017495.1 XM_017017496.1 XM_017017497.1 NM_001145308.4 NM_001145309.3 NM_001145310.3 XM_017017358.1

Gene Symbol
LRTOMT
Entrez Gene ID
220074
Gene Name
leucine rich transmembrane and O-methyltransferase domain containing
Gene Aliases
CFAP111, DFNB63, LRRC51
UniGene
Hs.317243
Gene Chromosome Location
Chr.11: 72079982-72110782 on Build GRCh38
Transcripts
NM_001278485.1 NM_001278486.1 NM_001278487.1 NM_001278488.1 NM_001278489.1 NM_001278490.1 NM_001278491.1 NM_001278492.1 NM_001278493.1 NM_001278494.1 NM_014042.2 XM_017017488.1 XM_017017489.1 XM_017017490.1 XM_017017491.1 XM_017017492.1 XM_017017493.1 XM_017017494.1 XM_017017495.1 XM_017017496.1 XM_017017497.1 NM_001145308.4 NM_001145309.3 NM_001145310.3 XM_017017358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6272797 c.181C>A p.H61N Chr.11: 72110225-72110225 on Build GRCh38
7293117 c.185A>G p.Y62C Chr.11: 72110221-72110221 on Build GRCh38
6317067 c.144C>A p.D48E Chr.11: 72110580-72110580 on Build GRCh38
4823006 c.132C>T p.I44I Chr.11: 72110592-72110592 on Build GRCh38
3967817 c.149A>G p.N50S Chr.11: 72110575-72110575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922112331 Chr.11: 72110637-72110637 on Build GRCh38
rs17849989 Chr.11: 72110550-72110550 on Build GRCh38
rs373529881 Chr.11: 72110506-72110506 on Build GRCh38
rs1014158498 Chr.11: 72110456-72110456 on Build GRCh38
rs1015612595 Chr.11: 72110659-72110659 on Build GRCh38
rs904996040 Chr.11: 72110247-72110247 on Build GRCh38
rs776177568 Chr.11: 72110259-72110259 on Build GRCh38
rs764832702 Chr.11: 72110280-72110281 on Build GRCh38
rs932261223 Chr.11: 72110667-72110667 on Build GRCh38
rs759869247 Chr.11: 72110594-72110594 on Build GRCh38
rs369798854 Chr.11: 72110496-72110496 on Build GRCh38
rs370124771 Chr.11: 72110480-72110480 on Build GRCh38
rs750009295 Chr.11: 72110508-72110509 on Build GRCh38
rs185280082 Chr.11: 72110293-72110293 on Build GRCh38
rs767267287 Chr.11: 72110697-72110697 on Build GRCh38
rs960727867 Chr.11: 72110329-72110329 on Build GRCh38
rs757509145 Chr.11: 72110641-72110641 on Build GRCh38
rs768617478 Chr.11: 72110564-72110564 on Build GRCh38
rs963425454 Chr.11: 72110567-72110567 on Build GRCh38
rs561436693 Chr.11: 72110254-72110254 on Build GRCh38
rs778349482 Chr.11: 72110253-72110253 on Build GRCh38
rs778261727 Chr.11: 72110517-72110517 on Build GRCh38
rs749197566 Chr.11: 72110558-72110558 on Build GRCh38
rs946481782 Chr.11: 72110530-72110530 on Build GRCh38
rs1015728484 Chr.11: 72110345-72110345 on Build GRCh38
rs200898905 Chr.11: 72110527-72110527 on Build GRCh38
rs202222531 Chr.11: 72110590-72110590 on Build GRCh38
rs138271743 Chr.11: 72110589-72110589 on Build GRCh38
rs886048639 Chr.11: 72110325-72110326 on Build GRCh38
rs532299898 Chr.11: 72110320-72110320 on Build GRCh38
rs370192741 Chr.11: 72110610-72110610 on Build GRCh38
rs974532751 Chr.11: 72110396-72110396 on Build GRCh38
rs781626202 Chr.11: 72110644-72110644 on Build GRCh38
rs945839322 Chr.11: 72110346-72110346 on Build GRCh38
rs745551454 Chr.11: 72110532-72110532 on Build GRCh38
rs759367241 Chr.11: 72110288-72110288 on Build GRCh38
rs759032367 Chr.11: 72110511-72110511 on Build GRCh38
rs753915114 Chr.11: 72110608-72110608 on Build GRCh38
rs962676481 Chr.11: 72110664-72110664 on Build GRCh38
rs905048896 Chr.11: 72110542-72110542 on Build GRCh38
rs904248125 Chr.11: 72110365-72110365 on Build GRCh38
rs539199138 Chr.11: 72110592-72110592 on Build GRCh38
rs375635711 Chr.11: 72110507-72110507 on Build GRCh38
rs561551303 Chr.11: 72110320-72110321 on Build GRCh38
rs894914936 Chr.11: 72110327-72110327 on Build GRCh38
rs770757770 Chr.11: 72110257-72110257 on Build GRCh38
rs767356574 Chr.11: 72110591-72110591 on Build GRCh38
rs995625058 Chr.11: 72110680-72110680 on Build GRCh38
rs894406121 Chr.11: 72110444-72110444 on Build GRCh38
rs543014197 Chr.11: 72110645-72110645 on Build GRCh38
rs763858916 Chr.11: 72110292-72110292 on Build GRCh38
rs932792064 Chr.11: 72110413-72110413 on Build GRCh38
rs987098150 Chr.11: 72110423-72110423 on Build GRCh38
rs769123440 Chr.11: 72110533-72110533 on Build GRCh38
rs772497729 Chr.11: 72110244-72110244 on Build GRCh38
rs766174504 Chr.11: 72110637-72110638 on Build GRCh38
rs557706099 Chr.11: 72110597-72110597 on Build GRCh38
rs199958250 Chr.11: 72110635-72110635 on Build GRCh38
rs201339132 Chr.11: 72110494-72110494 on Build GRCh38
rs754110853 Chr.11: 72110501-72110501 on Build GRCh38
rs536269368 Chr.11: 72110441-72110441 on Build GRCh38
rs554657170 Chr.11: 72110672-72110672 on Build GRCh38
rs766470230 Chr.11: 72110497-72110497 on Build GRCh38
rs568780022 Chr.11: 72110410-72110410 on Build GRCh38
rs747419504 Chr.11: 72110521-72110521 on Build GRCh38
rs757673616 Chr.11: 72110526-72110526 on Build GRCh38
rs764658753 Chr.11: 72110624-72110624 on Build GRCh38
rs546427194 Chr.11: 72110273-72110273 on Build GRCh38
rs896403111 Chr.11: 72110630-72110630 on Build GRCh38
rs561162900 Chr.11: 72110698-72110698 on Build GRCh38
rs912805774 Chr.11: 72110469-72110469 on Build GRCh38
rs962791943 Chr.11: 72110350-72110350 on Build GRCh38
rs192963404 Chr.11: 72110261-72110261 on Build GRCh38
rs758099655 Chr.11: 72110514-72110515 on Build GRCh38
rs982774430 Chr.11: 72110331-72110331 on Build GRCh38
rs769372273 Chr.11: 72110263-72110263 on Build GRCh38
rs550446274 Chr.11: 72110409-72110409 on Build GRCh38
rs913535244 Chr.11: 72110529-72110529 on Build GRCh38
rs752875020 Chr.11: 72110606-72110606 on Build GRCh38

View Full Product Details