Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72110609-72111117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC15
Entrez Gene ID
25906
Gene Name
anaphase promoting complex subunit 15
Gene Aliases
APC15, C11orf51, HSPC020
UniGene
Hs.38044
Gene Chromosome Location
Chr.11: 72106254-72112793 on Build GRCh38
Transcripts
NM_001278485.1 NM_001278486.1 NM_001278487.1 NM_001278488.1 NM_001278489.1 NM_001278490.1 NM_001278491.1 NM_001278492.1 NM_001278493.1 NM_001278494.1 NM_014042.2 XM_017017488.1 XM_017017489.1 XM_017017490.1 XM_017017491.1 XM_017017492.1 XM_017017493.1 XM_017017494.1 XM_017017495.1 XM_017017496.1 XM_017017497.1 NM_001145308.4 NM_001145309.3 NM_001145310.3 XM_017017358.1

Gene Symbol
LRTOMT
Entrez Gene ID
220074
Gene Name
leucine rich transmembrane and O-methyltransferase domain containing
Gene Aliases
CFAP111, DFNB63, LRRC51
UniGene
Hs.317243
Gene Chromosome Location
Chr.11: 72079982-72110782 on Build GRCh38
Transcripts
NM_001278485.1 NM_001278486.1 NM_001278487.1 NM_001278488.1 NM_001278489.1 NM_001278490.1 NM_001278491.1 NM_001278492.1 NM_001278493.1 NM_001278494.1 NM_014042.2 XM_017017488.1 XM_017017489.1 XM_017017490.1 XM_017017491.1 XM_017017492.1 XM_017017493.1 XM_017017494.1 XM_017017495.1 XM_017017496.1 XM_017017497.1 NM_001145308.4 NM_001145309.3 NM_001145310.3 XM_017017358.1

SNP Detail

SNP ID Chromosome Location
rs527413147 Chr.11: 72111029-72111029 on Build GRCh38
rs533733510 Chr.11: 72110915-72110915 on Build GRCh38
rs973507109 Chr.11: 72110967-72110967 on Build GRCh38
rs922112331 Chr.11: 72110637-72110637 on Build GRCh38
rs142914900 Chr.11: 72110735-72110735 on Build GRCh38
rs912690379 Chr.11: 72110704-72110704 on Build GRCh38
rs1015612595 Chr.11: 72110659-72110659 on Build GRCh38
rs566590072 Chr.11: 72110871-72110871 on Build GRCh38
rs1004897813 Chr.11: 72110878-72110878 on Build GRCh38
rs756082710 Chr.11: 72110797-72110797 on Build GRCh38
rs932261223 Chr.11: 72110667-72110667 on Build GRCh38
rs886048640 Chr.11: 72110713-72110713 on Build GRCh38
rs767267287 Chr.11: 72110697-72110697 on Build GRCh38
rs757509145 Chr.11: 72110641-72110641 on Build GRCh38
rs937900302 Chr.11: 72110801-72110801 on Build GRCh38
rs34775253 Chr.11: 72110887-72110888 on Build GRCh38
rs757088515 Chr.11: 72111033-72111033 on Build GRCh38
rs570532487 Chr.11: 72111061-72111061 on Build GRCh38
rs896289145 Chr.11: 72110928-72110928 on Build GRCh38
rs752561720 Chr.11: 72110730-72110730 on Build GRCh38
rs551610868 Chr.11: 72110783-72110783 on Build GRCh38
rs566343878 Chr.11: 72111022-72111022 on Build GRCh38
rs147422048 Chr.11: 72110789-72110789 on Build GRCh38
rs753511306 Chr.11: 72111026-72111026 on Build GRCh38
rs781626202 Chr.11: 72110644-72110644 on Build GRCh38
rs962676481 Chr.11: 72110664-72110664 on Build GRCh38
rs1056705901 Chr.11: 72110890-72110890 on Build GRCh38
rs1014917352 Chr.11: 72110883-72110883 on Build GRCh38
rs78756927 Chr.11: 72111012-72111012 on Build GRCh38
rs995625058 Chr.11: 72110680-72110680 on Build GRCh38
rs79122538 Chr.11: 72111050-72111050 on Build GRCh38
rs1004148840 Chr.11: 72110975-72110975 on Build GRCh38
rs543014197 Chr.11: 72110645-72110645 on Build GRCh38
rs543976625 Chr.11: 72110837-72110837 on Build GRCh38
rs184366262 Chr.11: 72111038-72111038 on Build GRCh38
rs766174504 Chr.11: 72110637-72110638 on Build GRCh38
rs1028444995 Chr.11: 72111030-72111030 on Build GRCh38
rs548212353 Chr.11: 72111015-72111015 on Build GRCh38
rs1024230325 Chr.11: 72110812-72110812 on Build GRCh38
rs954271952 Chr.11: 72111084-72111084 on Build GRCh38
rs199958250 Chr.11: 72110635-72110635 on Build GRCh38
rs190554032 Chr.11: 72110792-72110792 on Build GRCh38
rs573332460 Chr.11: 72110699-72110699 on Build GRCh38
rs926358901 Chr.11: 72110798-72110798 on Build GRCh38
rs554657170 Chr.11: 72110672-72110672 on Build GRCh38
rs193214519 Chr.11: 72110979-72110979 on Build GRCh38
rs561162900 Chr.11: 72110698-72110698 on Build GRCh38
rs879276724 Chr.11: 72110997-72110997 on Build GRCh38
rs886988911 Chr.11: 72110874-72110874 on Build GRCh38

View Full Product Details